Assay Details
| Gene Symbol: Ccbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172404.2 | NP_765992.2 | 7 - 8 | 840 | 62 | |
| GenBank mRNA | AK008165.1 | - | 5 - 6 | 617 | 62 | |
| AK031511.1 | - | 7 - 8 | 853 | 62 | ||
| AK042391.1 | - | 6 - 7 | 786 | 62 | ||
| AK088404.1 | - | 6 - 7 | 748 | 62 | ||
| AK163165.1 | - | 6 - 7 | 648 | 62 | ||
| AK218327.1 | - | 1 - 2 | 84 | 62 | ||
| BC016206.1 | - | 7 - 8 | 942 | 4912335 | 62 | |
| BC025124.1 | - | 6 - 7 | 973 | 4486058 | 62 | |
| BC052047.1 | - | 7 - 8 | 840 | 5687475 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccbl1
Genomic Map