Assay Details
Gene Symbol: Tmx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029148.1 | NP_083424.1 | 3 - 4 | 443 | 59 | |
GenBank mRNA | AK030696.1 | - | 3 - 4 | 457 | 59 | |
AK033111.1 | - | 3 - 4 | 365 | 59 | ||
AK040779.1 | - | 3 - 4 | 428 | 59 | ||
AK053897.1 | - | 3 - 4 | 2574 | 59 | ||
AK129301.1 | - | 3 - 4 | 442 | 59 | ||
AK147465.1 | - | 3 - 4 | 444 | 59 | ||
BC050918.1 | - | 3 - 4 | 342 | 59 | ||
BC054558.1 | - | 3 - 4 | 443 | 6405599 | 59 | |
BC138168.1 | - | 3 - 4 | 417 | 8861187 | 59 | |
BC138169.1 | - | 3 - 4 | 417 | 8861188 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
