Assay Details
Gene Symbol: Tmx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029148.1 | NP_083424.1 | 4 - 5 | 568 | 85 | |
GenBank mRNA | AK015667.1 | - | 4 - 5 | 581 | 85 | |
AK030696.1 | - | 4 - 5 | 582 | 85 | ||
AK040779.1 | - | 4 - 5 | 553 | 85 | ||
AK053897.1 | - | 4 - 5 | 2699 | 85 | ||
AK129301.1 | - | 4 - 5 | 567 | 85 | ||
AK147465.1 | - | 4 - 5 | 569 | 85 | ||
BC050918.1 | - | 4 - 5 | 467 | 85 | ||
BC054558.1 | - | 4 - 5 | 568 | 6405599 | 85 | |
BC138168.1 | - | 4 - 5 | 542 | 8861187 | 85 | |
BC138169.1 | - | 4 - 5 | 542 | 8861188 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
