Assay Details
| Gene Symbol: Wdr25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177602.3 | NP_808270.2 | 4 - 5 | 1138 | 62 | |
| GenBank mRNA | AK147993.1 | - | 5 - 6 | 2840 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdr25
Genomic Map