Assay Details
| Gene Symbol: Dgkz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166597.1 | NP_001160069.1 | 12 - 13 | 1732 | 90 | |
| NM_138306.2 | NP_612179.2 | 11 - 12 | 1131 | 90 | ||
| GenBank mRNA | AK087503.1 | - | 12 - 13 | 1733 | 90 | |
| AK170936.1 | - | 8 - 9 | 754 | 90 | ||
| BC049228.1 | - | 11 - 12 | 1167 | 3668704 | 90 | |
| BQ443200.1 | - | 2 - 3 | 135 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dgkz
Genomic Map