Assay Details
Gene Symbol: Obp2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099301.1 | NP_001092771.1 | 4 - 5 | 450 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1986476 | NM_001099301.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 533582 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211125 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 47693 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1986476 | NM_001099301.1 | mouse_transcriptome |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074022 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074002 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073976 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074025 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073975 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073982 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073964 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21074011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073980 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21073979 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21074032 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01477366 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21074007 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01780989 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01477371 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21074025 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01477322 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21073982 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21074027 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01899168 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21073984 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01477315 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01477356 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21074031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21074015 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01477339 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01477329 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01477361 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21074013 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21073987 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21074009 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01477311 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21073967 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01780990 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21073989 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21073976 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01477364 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01477355 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21074022 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01477354 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21074023 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10470187 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470188 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470191 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470190 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10470189 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10470186 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 304194 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1234176 | - | - |
