Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700019734.mm.1 |
TC0700002163.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019734.mm.1 |
NM_207270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019734.mm.1 |
NM_207270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019735.mm.1 |
TC0700002163.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019735.mm.1 |
NM_207270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019735.mm.1 |
NM_207270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019737.mm.1 |
TC0700002163.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019737.mm.1 |
NM_207270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019737.mm.1 |
NM_207270.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019736.mm.1 |
TC0700002163.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700019736.mm.1 |
NM_207270.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700019736.mm.1 |
NM_207270.2 |
Refseq |