Assay Details
Gene Symbol: Ptprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207270.2 | NP_997153.2 | 15 - 16 | 2562 | 77 | |
GenBank mRNA | AB217856.1 | - | 14 - 15 | 2235 | 77 | |
AK078884.1 | - | 14 - 15 | 2239 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
