Assay Details
Gene Symbol: Ptprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207270.2 | NP_997153.2 | 16 - 17 | 2701 | 68 | |
GenBank mRNA | AB217856.1 | - | 15 - 16 | 2374 | 68 | |
AK078884.1 | - | 15 - 16 | 2378 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130449 | AB217856.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130449 | AK078884.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P130449 | NM_207270.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 57072 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 601493 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830355 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 772048 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 230397 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918934 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928383 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46200 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 377332 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 193301 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98967 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770148 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 471346 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 745671 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783717 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300817 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30069 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 134493 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38536 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159637 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P103899 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102077 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130449 | AB217856.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130449 | AK078884.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P130449 | NM_207270.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P103899 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06114013 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06114036 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01475648 | - | - |
Whole Mouse Genome G4122A | A_51_P365666 | AB217856.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P365666 | AK078884.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P365666 | NM_207270.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P103899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949347 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949414 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949431 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949570 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949429 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949469 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949494 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949491 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949465 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949409 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949412 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949391 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949595 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949454 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949445 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949437 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949363 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949584 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949355 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949407 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949471 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949572 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949488 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949617 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949381 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949551 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949578 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949443 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949590 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949371 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949503 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949478 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949486 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949458 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949547 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949487 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949540 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949410 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949535 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949563 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949474 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949434 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949388 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949370 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949361 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949418 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949373 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949398 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949415 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949580 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949462 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949350 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949426 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949558 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949484 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949440 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949427 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949376 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949405 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949450 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949464 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949430 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24949369 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949468 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949540 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06031511 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949426 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06559363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949474 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06394868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949571 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949545 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06058117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949417 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949554 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06422375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949578 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949473 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949573 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949477 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949582 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06487678 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949585 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949404 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06422376 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06516443 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949470 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949398 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949617 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949407 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949412 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06123158 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06249832 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949465 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949405 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949410 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949551 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949419 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949544 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949563 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06249828 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949421 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06427907 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06568323 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06623395 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949560 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949427 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949570 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949401 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949555 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05885936 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06289449 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949469 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949423 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949409 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949583 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06394870 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949535 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06390389 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06351771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949550 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949414 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06197079 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949558 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949478 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24949588 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10559599 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559592 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559591 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559594 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559601 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559598 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559595 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559600 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559604 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559603 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559602 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559597 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559596 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559605 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10559593 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10559590 | - | - |
Affymetrix Mouse Clariom S Array | PSR0700019731.mm.1 | TC0700002163.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700019731.mm.1 | NM_207270.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700019731.mm.1 | NM_207270.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700019732.mm.1 | TC0700002163.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700019732.mm.1 | NM_207270.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700019732.mm.1 | NM_207270.2 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 588688 | - | - |
ABI Mouse Genome Survey Microarray | 635956 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3870070 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3870070 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003870070 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2753732 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2753732 | - | - |
mouse-6 v1.1 (Illumina) | scl0399642.2_8-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3870070 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0399642.2_8-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3870070 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1253963 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1253963 | - | - |
