Assay Details
| Gene Symbol: Dtnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025772.4 | NP_080048.2 | 1 - 2 | 187 | 73 | |
| GenBank mRNA | AJ404859.1 | - | 1 - 2 | 186 | 73 | |
| AK150306.1 | - | 1 - 2 | 183 | 73 | ||
| AK159656.1 | - | 1 - 2 | 312 | 73 | ||
| BC018350.1 | - | 1 - 2 | 178 | 4195234 | 73 | |
| BC048682.1 | - | 1 - 2 | 232 | 73 | ||
| BC058574.1 | - | 1 - 2 | 160 | 6487720 | 73 | |
More Information
Set Membership: |
|

Dtnbp1
Genomic Map