Assay Details
| Gene Symbol: Igf2bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023670.3 | NP_076159.3 | 3 - 4 | 775 | 71 | |
| GenBank mRNA | AB046173.1 | - | 3 - 4 | 292 | 71 | |
| AK011689.1 | - | 3 - 4 | 556 | 71 | ||
| AK030096.1 | - | 3 - 4 | 635 | 71 | ||
| AK088465.1 | - | 3 - 4 | 534 | 71 | ||
| AK147140.1 | - | 3 - 4 | 779 | 71 | ||
| BC045138.1 | - | 3 - 4 | 405 | 6306448 | 71 | |
| BC049082.1 | - | 3 - 4 | 441 | 5706238 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igf2bp3
Genomic Map