Assay Details
| Gene Symbol: Igf2bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023670.3 | NP_076159.3 | 4 - 5 | 825 | 94 | |
| GenBank mRNA | AB046173.1 | - | 4 - 5 | 342 | 94 | |
| AK011689.1 | - | 4 - 5 | 606 | 94 | ||
| AK030096.1 | - | 4 - 5 | 685 | 94 | ||
| AK088465.1 | - | 4 - 5 | 584 | 94 | ||
| AK147140.1 | - | 4 - 5 | 829 | 94 | ||
| BC045138.1 | - | 4 - 5 | 455 | 6306448 | 94 | |
| BC049082.1 | - | 4 - 5 | 491 | 5706238 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Igf2bp3
Genomic Map