Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600017599.mm.1 |
TC0600002240.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017599.mm.1 |
NM_198102.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017599.mm.1 |
NM_198102.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017598.mm.1 |
TC0600002240.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017598.mm.1 |
NM_198102.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017598.mm.1 |
NM_198102.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017597.mm.1 |
TC0600002240.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600017597.mm.1 |
NM_198102.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600017597.mm.1 |
NM_198102.2 |
Refseq |