Assay Details
Gene Symbol: Tra2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198102.2 | NP_932770.2 | 7 - 8 | 1017 | 65 | |
GenBank mRNA | AK089980.1 | - | 8 - 9 | 998 | 65 | |
AK164994.1 | - | 2 - 3 | 158 | 65 | ||
AK171708.1 | - | 6 - 7 | 2237 | 65 | ||
BC023018.1 | - | 4 - 5 | 397 | 65 | ||
BC057448.1 | - | 7 - 8 | 972 | 4016620 | 65 | |
BC058764.1 | - | 8 - 9 | 1282 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431875 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431871 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431873 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431878 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431870 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431874 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431879 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431872 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431876 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513163 | BC023018.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513163 | BC057448.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513163 | BC058764.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513163 | BE134105.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513163 | AK171708.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513163 | BC023018.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513163 | BC057448.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513163 | BC058764.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513163 | BE134105.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P513163 | AK171708.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649492 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 523420 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456919 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 722383 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 420 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 636210 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 953691 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249683 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 136674 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 232599 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450991 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 961999 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 85004 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 170146 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133989 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137697 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513163 | BC023018.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513163 | BC057448.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513163 | BC058764.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513163 | BE134105.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513163 | AK171708.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513163 | BC023018.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513163 | BC057448.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513163 | BC058764.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513163 | BE134105.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P513163 | AK171708.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513163 | BC023018.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513163 | BC057448.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513163 | BC058764.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513163 | BE134105.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513163 | AK171708.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513163 | BC023018.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513163 | BC057448.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513163 | BC058764.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513163 | BE134105.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P513163 | AK171708.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05892222 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137697 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05892230 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133989 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05892240 | - | - |
Whole Mouse Genome G4122A | A_51_P513163 | BC023018.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513163 | BC057448.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513163 | BC058764.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513163 | BE134105.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513163 | AK171708.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513163 | BC023018.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513163 | BC057448.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513163 | BC058764.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513163 | BE134105.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P513163 | AK171708.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P591431 | BC057448.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P591431 | BC058764.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P591431 | AK171708.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P591431 | AK089980.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P591431 | NM_198102.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133989 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137697 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05254997 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05573138 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05254986 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431876 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431871 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431878 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431873 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431875 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431870 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431874 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431879 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24431872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431770 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24431831 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431884 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431835 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431812 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431863 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05713386 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05853735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05307470 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431858 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05646167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431809 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05823880 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431881 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05840536 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431880 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431806 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431855 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05840537 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431899 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431888 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431882 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431896 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431849 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431801 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431830 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05450008 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431879 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431870 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431854 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431848 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05254997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05487167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05255001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431865 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431832 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431900 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431798 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431841 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05254994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05465257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05334783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431866 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431902 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431886 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431818 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431816 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05254986 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431860 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431803 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431808 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431862 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05207185 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431802 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431805 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05254990 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05374342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05048294 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431828 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24431846 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10544635 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544632 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544639 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544637 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544633 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544636 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544634 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544630 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10544631 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1439043_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1454725_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429680_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10544629 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10544638 | - | - |
Mouse Genome 430 2.0 Array | 1454725_at | - | - |
Mouse Genome 430 2.0 Array | 1454725_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101214_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101214_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600017594.mm.1 | TC0600002240.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600017594.mm.1 | NM_198102.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600017594.mm.1 | NM_198102.2 | Refseq |
Affymetrix HT Mouse Genome 430A Array | 1454725_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1454725_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1439043_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429680_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1439043_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429680_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6860446 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006860446 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2687454 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2687454 | - | - |
mouse-6 v1.1 (Illumina) | scl0101214.1_105-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6860446 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0101214.1_105-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6860446 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2857086 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2857086 | - | - |
