Assay Details
| Gene Symbol: Gas7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF133184.1 | - | 6 - 7 | 826 | 58 | |
| AK154463.1 | - | 10 - 11 | 951 | 58 | ||
| AK172943.1 | - | 6 - 7 | 770 | 58 | ||
| BC120528.1 | - | 10 - 11 | 875 | 40129451 | 58 | |
| BC120530.1 | - | 10 - 11 | 875 | 40129453 | 58 | |
| U19860.1 | - | 10 - 11 | 980 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gas7
Genomic Map