Assay Details
| Gene Symbol: Gas7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109657.3 | NP_001103127.1 | 11 - 12 | 1271 | 57 | |
| NM_001277079.1 | NP_001264008.1 | 11 - 12 | 1100 | 57 | ||
| NM_001277080.1 | NP_001264009.1 | 10 - 11 | 1014 | 57 | ||
| NM_008088.5 | NP_032114.2 | 11 - 12 | 1077 | 57 | ||
| GenBank mRNA | AF133184.1 | - | 7 - 8 | 949 | 57 | |
| AK017394.1 | - | 11 - 12 | 1070 | 57 | ||
| AK076926.1 | - | 12 - 13 | 1153 | 57 | ||
| AK152149.1 | - | 9 - 10 | 774 | 57 | ||
| AK154463.1 | - | 11 - 12 | 1074 | 57 | ||
| AK172943.1 | - | 7 - 8 | 893 | 57 | ||
| BC120528.1 | - | 11 - 12 | 998 | 40129451 | 57 | |
| BC120530.1 | - | 11 - 12 | 998 | 40129453 | 57 | |
| CV562268.1 | - | 3 - 4 | 315 | 57 | ||
| U19860.1 | - | 11 - 12 | 1103 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gas7
Genomic Map