Assay Details
Gene Symbol: Usp43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291049.1 | NP_001277978.1 | 1 - 2 | 610 | 65 | |
NM_173754.4 | NP_776115.2 | 1 - 2 | 610 | 65 | ||
GenBank mRNA | AK042217.1 | - | 1 - 2 | 610 | 65 | |
AK083271.1 | - | 1 - 2 | 448 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
