Assay Details
| Gene Symbol: Exd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172857.2 | NP_766445.1 | 9 - 10 | 1404 | 91 | |
| GenBank mRNA | AK030132.1 | - | 9 - 10 | 1406 | 91 | |
| BC092241.1 | - | 7 - 8 | 658 | 30670567 | 91 | |
| BC144750.1 | - | 8 - 9 | 787 | 9053298 | 91 | |
| BC150662.1 | - | 9 - 10 | 881 | 9087573 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Exd1
Genomic Map