Assay Details
Gene Symbol: Tlk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172664.3 | NP_766252.2 | 2 - 3 | 406 | 113 | |
GenBank mRNA | AK029773.1 | - | 1 - 2 | 132 | 113 | |
BC051641.1 | - | 2 - 3 | 406 | 6507968 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
