Assay Details
Gene Symbol: Tlk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172664.3 | NP_766252.2 | 9 - 10 | 988 | 153 | |
GenBank mRNA | AK029773.1 | - | 8 - 9 | 714 | 153 | |
AK172897.1 | - | 3 - 4 | 252 | 153 | ||
BC033931.1 | - | 7 - 8 | 550 | 153 | ||
BC051641.1 | - | 9 - 10 | 988 | 6507968 | 153 | |
BC057369.1 | - | 10 - 11 | 974 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
