Assay Details
| Gene Symbol: Nipa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001256130.1 | NP_001243059.1 | 1 - 2 | 294 | 100 | |
| GenBank mRNA | AB041581.1 | - | 1 - 2 | 231 | 100 | |
| AK008106.1 | - | 1 - 2 | 280 | 100 | ||
| AK031255.1 | - | 1 - 2 | 259 | 100 | ||
| AK041427.1 | - | 1 - 2 | 295 | 100 | ||
| AK154755.1 | - | 1 - 2 | 252 | 100 | ||
| BC038499.1 | - | 1 - 2 | 236 | 4482150 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB041581.1, AK041427.1, AK154755.1, BC038499.1, NM_001256130.1 transcripts.
Set Membership: |
|

Nipa2
Genomic Map