Assay Details
| Gene Symbol: Scn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018733.2 | NP_061203.2 | 19 - 20 | 3858 | 97 | |
| GenBank mRNA | EF591872.1 | - | 3 - 4 | 663 | 97 | |
| L42339.1 | - | 3 - 4 | 611 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scn1a
Genomic Map