Assay Details
Gene Symbol: Galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015736.2 | NP_056551.2 | 5 - 6 | 1445 | 74 | |
GenBank mRNA | AK019995.1 | - | 5 - 6 | 1444 | 74 | |
AK135489.1 | - | 5 - 6 | 1406 | 74 | ||
BC043331.1 | - | 5 - 6 | 1301 | 5342768 | 74 | |
U70538.1 | - | 5 - 6 | 1289 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
