Assay Details
| Gene Symbol: Galnt3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015736.2 | NP_056551.2 | 7 - 8 | 1764 | 129 | |
| GenBank mRNA | AK019995.1 | - | 7 - 8 | 1763 | 129 | |
| AK135489.1 | - | 7 - 8 | 1725 | 129 | ||
| BC043331.1 | - | 7 - 8 | 1620 | 5342768 | 129 | |
| U70538.1 | - | 7 - 8 | 1608 | 129 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Galnt3
Genomic Map