Assay Details
Gene Symbol: Galnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015736.2 | NP_056551.2 | 8 - 9 | 1907 | 79 | |
GenBank mRNA | AK019995.1 | - | 8 - 9 | 1906 | 79 | |
AK135489.1 | - | 8 - 9 | 1868 | 79 | ||
BC043331.1 | - | 8 - 9 | 1763 | 5342768 | 79 | |
U70538.1 | - | 8 - 9 | 1751 | 79 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
