Assay Details
| Gene Symbol: Sco1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040026.1 | NP_001035115.1 | 4 - 5 | 623 | 69 | |
| GenBank mRNA | AK011251.1 | - | 4 - 5 | 639 | 69 | |
| AK017794.1 | - | 4 - 5 | 624 | 69 | ||
| AK139512.1 | - | 4 - 5 | 594 | 69 | ||
| AK167305.1 | - | 4 - 5 | 488 | 69 | ||
| BC139008.1 | - | 4 - 5 | 618 | 8862030 | 69 | |
| BC139009.1 | - | 4 - 5 | 618 | 8862031 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sco1
Genomic Map