Assay Details
Gene Symbol: Kcnh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133207.2 | NP_573470.2 | 6 - 7 | 1339 | 72 | |
GenBank mRNA | AJ291608.1 | - | 6 - 7 | 1146 | 72 | |
AK034003.1 | - | 6 - 7 | 1340 | 72 | ||
AK052366.1 | - | 5 - 6 | 1202 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
