Assay Details
Gene Symbol: Kcnh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133207.2 | NP_573470.2 | 8 - 9 | 2164 | 153 | |
GenBank mRNA | AJ291608.1 | - | 8 - 9 | 1971 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
