Assay Details
Gene Symbol: Pdss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019501.3 | NP_062374.2 | 2 - 3 | 309 | 69 | |
GenBank mRNA | AB210841.1 | - | 2 - 3 | 229 | 69 | |
AK012307.1 | - | 2 - 3 | 237 | 69 | ||
BC026820.1 | - | 2 - 3 | 211 | 4039464 | 69 | |
BC107273.1 | - | 2 - 3 | 124 | 69 | ||
BC107274.1 | - | 2 - 3 | 128 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC026820.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
