Assay Details
Gene Symbol: Glp2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175681.3 | NP_783612.2 | 1 - 2 | 343 | 87 | |
GenBank mRNA | AY605231.1 | - | 1 - 2 | 62 | 87 | |
BC044746.2 | - | 1 - 2 | 343 | 5363416 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
