Assay Details
| Gene Symbol: Glp2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175681.3 | NP_783612.2 | 3 - 4 | 545 | 70 | |
| GenBank mRNA | AY255567.1 | - | 2 - 3 | 159 | 70 | |
| AY605231.1 | - | 3 - 4 | 264 | 70 | ||
| BC044746.2 | - | 3 - 4 | 545 | 5363416 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glp2r
Genomic Map