Assay Details
Gene Symbol: Myh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177369.3 | NP_796343.2 | 20 - 21 | 2475 | 81 | |
GenBank mRNA | AK029328.1 | - | 20 - 21 | 2386 | 81 | |
BC150737.1 | - | 20 - 21 | 2398 | 9087648 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
