Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200016015.mm.1 |
NM_153501.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016015.mm.1 |
NM_153501.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016015.mm.1 |
TC0200002013.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016021.mm.1 |
NM_153501.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016021.mm.1 |
NM_153501.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016021.mm.1 |
TC0200002013.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016020.mm.1 |
NM_153501.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016020.mm.1 |
NM_153501.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016020.mm.1 |
TC0200002013.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200016012.mm.1 |
NM_153501.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016012.mm.1 |
NM_153501.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200016012.mm.1 |
TC0200002013.mm.1 |
MTA TC |