Assay Details
| Gene Symbol: Ptpra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163688.1 | NP_001157160.1 | 6 - 7 | 1002 | 74 | |
| NM_008980.2 | NP_033006.2 | 6 - 7 | 1002 | 74 | ||
| GenBank mRNA | AF332081.1 | - | 5 - 6 | 715 | 74 | |
| AF332082.1 | - | 5 - 6 | 715 | 74 | ||
| AJ431367.1 | - | 5 - 6 | 715 | 74 | ||
| AK010458.1 | - | 6 - 7 | 1004 | 74 | ||
| AK031690.1 | - | 6 - 7 | 1001 | 74 | ||
| AK165673.1 | - | 6 - 7 | 1000 | 74 | ||
| BC050930.1 | - | 6 - 7 | 984 | 6811172 | 74 | |
| BC110683.1 | - | 6 - 7 | 968 | 3965311 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptpra
Genomic Map