Assay Details
Gene Symbol: Hspa12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028306.3 | NP_082582.1 | 3 - 4 | 260 | 85 | |
GenBank mRNA | AK012548.1 | - | 3 - 4 | 233 | 85 | |
AY196790.1 | - | 2 - 3 | 140 | 85 | ||
BC011103.1 | - | 3 - 4 | 260 | 3990088 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
