Assay Details
| Gene Symbol: Cdc25b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001111075.4 | NP_001104545.1 | 13 - 14 | 1637 | 139 | |
| NM_023117.4 | NP_075606.1 | 14 - 15 | 1715 | 139 | ||
| GenBank mRNA | AK004836.1 | - | 13 - 14 | 1594 | 139 | |
| AK043730.1 | - | 13 - 14 | 1628 | 139 | ||
| AK153903.1 | - | 14 - 15 | 1716 | 139 | ||
| AK157397.1 | - | 13 - 14 | 1595 | 139 | ||
| BC002287.1 | - | 5 - 6 | 502 | 139 | ||
| BC057568.1 | - | 13 - 14 | 1508 | 6401489 | 139 | |
| S93521.1 | - | 14 - 15 | 1584 | 139 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdc25b
Genomic Map