Assay Details
| Gene Symbol: Epb4.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013513.2 | NP_038541.1 | 11 - 12 | 1939 | 71 | |
| GenBank mRNA | AK080230.1 | - | 2 - 3 | 225 | 71 | |
| AK089080.1 | - | 5 - 6 | 987 | 71 | ||
| AK133896.1 | - | 11 - 12 | 1953 | 71 | ||
| AK143841.1 | - | 11 - 12 | 1919 | 71 | ||
| AK158219.1 | - | 11 - 12 | 1857 | 71 | ||
| AK158247.1 | - | 11 - 12 | 1903 | 71 | ||
| AK165288.1 | - | 11 - 12 | 1866 | 71 | ||
| AK172410.1 | - | 11 - 12 | 1903 | 71 | ||
| BC053739.1 | - | 11 - 12 | 1911 | 71 | ||
| BC066194.1 | - | 11 - 12 | 1939 | 30429971 | 71 | |
| BC066806.1 | - | 11 - 12 | 1939 | 30429961 | 71 | |
| L35933.1 | - | 11 - 12 | 1918 | 71 | ||
| U03487.1 | - | 11 - 12 | 1869 | 71 | ||
| U04055.1 | - | 11 - 12 | 1939 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epb4.2
Genomic Map