Assay Details
| Gene Symbol: Epb4.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013513.2 | NP_038541.1 | 8 - 9 | 1236 | 55 | |
| GenBank mRNA | AK089080.1 | - | 2 - 3 | 284 | 55 | |
| AK133896.1 | - | 8 - 9 | 1250 | 55 | ||
| AK137484.1 | - | 8 - 9 | 1317 | 55 | ||
| AK143841.1 | - | 8 - 9 | 1216 | 55 | ||
| AK158219.1 | - | 8 - 9 | 1154 | 55 | ||
| AK158247.1 | - | 8 - 9 | 1200 | 55 | ||
| AK158370.1 | - | 8 - 9 | 1163 | 55 | ||
| AK165288.1 | - | 8 - 9 | 1163 | 55 | ||
| AK172410.1 | - | 8 - 9 | 1200 | 55 | ||
| BC053739.1 | - | 8 - 9 | 1208 | 55 | ||
| L35933.1 | - | 8 - 9 | 1215 | 55 | ||
| U04055.1 | - | 8 - 9 | 1236 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epb4.2
Genomic Map