Assay Details
| Gene Symbol: Pdia3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007952.2 | NP_031978.2 | 6 - 7 | 850 | 105 | |
| GenBank mRNA | AK088721.1 | - | 6 - 7 | 810 | 105 | |
| AK132459.1 | - | 6 - 7 | 815 | 105 | ||
| AK133799.1 | - | 6 - 7 | 809 | 105 | ||
| AK167807.1 | - | 6 - 7 | 803 | 105 | ||
| AK169611.1 | - | 6 - 7 | 804 | 105 | ||
| BC003285.1 | - | 6 - 7 | 769 | 3582199 | 105 | |
| BC033439.1 | - | 6 - 7 | 744 | 4016310 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdia3
Genomic Map