Assay Details
| Gene Symbol: Adal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290811.1 | NP_001277740.1 | 2 - 3 | 450 | 94 | |
| NM_001290812.1 | NP_001277741.1 | 2 - 3 | 393 | 94 | ||
| GenBank mRNA | AK016299.1 | - | 2 - 3 | 447 | 94 | |
| AK039677.1 | - | 2 - 3 | 383 | 94 | ||
| AK049697.1 | - | 2 - 3 | 443 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK016299.1, AK049697.1, NM_001290811.1, NM_001290812.1 transcripts.
Set Membership: |
|

Adal
Genomic Map