Assay Details
| Gene Symbol: Adal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290811.1 | NP_001277740.1 | 7 - 8 | 888 | 76 | |
| NM_001290812.1 | NP_001277741.1 | 5 - 6 | 674 | 76 | ||
| NM_029475.2 | NP_083751.1 | 7 - 8 | 780 | 76 | ||
| GenBank mRNA | AK016299.1 | - | 7 - 8 | 885 | 76 | |
| AK039677.1 | - | 5 - 6 | 664 | 76 | ||
| AK049697.1 | - | 7 - 8 | 881 | 76 | ||
| BC050879.1 | - | 7 - 8 | 732 | 5687835 | 76 | |
| BC052048.1 | - | 7 - 8 | 732 | 5687835 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adal
Genomic Map