Assay Details
Gene Symbol: Cacng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133190.1 | NP_573453.1 | 1 - 2 | 388 | 59 | |
GenBank mRNA | AF361350.1 | - | 1 - 2 | 388 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
