Assay Details
| Gene Symbol: Mboat7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029934.3 | NP_084210.2 | 5 - 6 | 1285 | 84 | |
| GenBank mRNA | AB055410.1 | - | 5 - 6 | 1283 | 84 | |
| AK019981.1 | - | 5 - 6 | 696 | 84 | ||
| AK038998.1 | - | 5 - 6 | 707 | 84 | ||
| AK044149.1 | - | 5 - 6 | 712 | 84 | ||
| AK150010.1 | - | 5 - 6 | 679 | 84 | ||
| AK154606.1 | - | 5 - 6 | 1284 | 84 | ||
| AK158998.1 | - | 4 - 5 | 1040 | 84 | ||
| AK170124.1 | - | 5 - 6 | 762 | 84 | ||
| BC023417.1 | - | 5 - 6 | 733 | 5028759 | 84 | |
| BC118958.2 | - | 5 - 6 | 930 | 40093821 | 84 | |
| EU016380.1 | - | 4 - 5 | 497 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mboat7
Genomic Map