Assay Details
Gene Symbol: Ifi27l2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281830.1 | NP_001268759.1 | 1 - 2 | 50 | 67 | |
NM_029803.3 | NP_084079.1 | 1 - 2 | 50 | 67 | ||
GenBank mRNA | AK040279.1 | - | 1 - 2 | 31 | 67 | |
AK160503.1 | - | 1 - 2 | 31 | 67 | ||
BC086889.1 | - | 1 - 2 | 30 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK160503.1, NM_001281830.1, NM_029803.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P90363 | NM_029803.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P90363 | AK160503.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P90363 | BC086889.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P90363 | BC100588.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P90363 | NM_029803.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P90363 | AK160503.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P90363 | BC086889.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P90363 | BC100588.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632957 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 387437 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P117197 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154580 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P175274 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P90363 | NM_029803.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P90363 | AK160503.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P90363 | BC086889.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P90363 | BC100588.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P90363 | NM_029803.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P90363 | AK160503.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P90363 | BC086889.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P90363 | BC100588.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697636 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697624 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697619 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697638 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697631 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697626 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697656 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697621 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697646 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697627 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697635 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697640 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697651 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697649 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697632 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697643 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697623 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697622 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697641 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697654 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697618 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697647 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697625 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28697645 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P117197 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154580 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P175274 | - | - |
Whole Mouse Genome G4122A | A_51_P358233 | AK010014.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P90363 | NM_029803.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P90363 | AK160503.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P90363 | BC086889.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P90363 | BC100588.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P90363 | NM_029803.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P90363 | AK160503.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P90363 | BC086889.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P90363 | BC100588.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P90364 | NM_029803.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P90364 | AK040279.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P90364 | AK160503.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P90364 | AY090098.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P90364 | BC086889.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108917 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P175274 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154580 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P117197 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09861233 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10118163 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10118173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697556 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697632 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697663 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697587 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697560 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697666 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697669 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697576 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697660 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697641 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697688 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697673 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697579 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697668 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697624 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697649 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28697622 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10369364 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697652 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697659 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697630 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697660 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10118157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10118170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697640 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09805691 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697638 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697668 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697621 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697666 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697622 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697649 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697651 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697672 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697665 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697654 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697627 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10349919 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10118159 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10118158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10118173 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697663 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697661 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10118151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697616 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697619 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697636 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10349924 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28697647 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10349915 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10402351 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402349 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402348 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402350 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402352 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1426278_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10402347 | - | - |
Mouse Genome 430 2.0 Array | 1426278_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1426278_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76933_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76933_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1200015762.mm.1 | NM_001281830.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200015762.mm.1 | NM_029803.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200015762.mm.1 | TC1200002269.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200015759.mm.1 | NM_029803.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200015759.mm.1 | TC1200002269.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200015757.mm.1 | NM_001281830.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200015757.mm.1 | TC1200002269.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200015760.mm.1 | NM_001281830.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200015760.mm.1 | NM_029803.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200015760.mm.1 | TC1200002269.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200015761.mm.1 | NM_001281830.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200015761.mm.1 | NM_029803.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200015761.mm.1 | TC1200002269.mm.1 | MTA TC |
Mouse Expression Set 430 | 1426278_at | - | - |
Mouse Expression Set 430 | 1426278_PM_at | - | - |
Murine Genome U74v2 Set | 92718_PM_at | - | - |
Murine Genome U74v2 Set | 92718_at | - | - |
Mouse Genome 430A 2.0 Array | 1426278_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1426278_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426278_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426278_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2340093 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2340093 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 100520438 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002340093 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2762944 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2762944 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2416594 | - | - |
mouse-6 v1.1 (Illumina) | scl42111.5.1_11-S | - | - |
mouse-6 v1.1 (Illumina) | scl0076933.2_252-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2340093 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 100520438 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0076933.2_252-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2340093 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 100520438 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2762944 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2762944 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0076933.2_252-S | - | - |
