Assay Details
| Gene Symbol: Ubr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025666.2 | NP_079942.1 | 6 - 7 | 655 | 73 | |
| GenBank mRNA | AK011968.1 | - | 6 - 7 | 526 | 73 | |
| AK017542.1 | - | 6 - 7 | 611 | 73 | ||
| AK088208.1 | - | 6 - 7 | 657 | 73 | ||
| AK146308.1 | - | 6 - 7 | 657 | 73 | ||
| AK149989.1 | - | 6 - 7 | 610 | 73 | ||
| AK155611.1 | - | 6 - 7 | 526 | 73 | ||
| BC051678.1 | - | 3 - 4 | 183 | 73 | ||
| BC058535.1 | - | 5 - 6 | 339 | 73 | ||
| BC079531.1 | - | 5 - 6 | 352 | 73 | ||
| BC094323.1 | - | 6 - 7 | 662 | 73 | ||
| BC094423.1 | - | 6 - 7 | 613 | 6812389 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubr7
Genomic Map