Assay Details
| Gene Symbol: Ubr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025666.2 | NP_079942.1 | 7 - 8 | 865 | 101 | |
| GenBank mRNA | AK088208.1 | - | 7 - 8 | 867 | 101 | |
| AK146308.1 | - | 7 - 8 | 867 | 101 | ||
| AK149989.1 | - | 7 - 8 | 820 | 101 | ||
| AK155611.1 | - | 7 - 8 | 736 | 101 | ||
| AK203922.1 | - | 2 - 3 | 228 | 101 | ||
| BC051678.1 | - | 4 - 5 | 393 | 101 | ||
| BC058535.1 | - | 6 - 7 | 549 | 101 | ||
| BC079531.1 | - | 6 - 7 | 562 | 101 | ||
| BC094323.1 | - | 7 - 8 | 872 | 101 | ||
| BC094423.1 | - | 7 - 8 | 823 | 6812389 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubr7
Genomic Map