Assay Details
| Gene Symbol: Ubr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025666.2 | NP_079942.1 | 8 - 9 | 1008 | 71 | |
| GenBank mRNA | AK088208.1 | - | 8 - 9 | 1010 | 71 | |
| AK146308.1 | - | 8 - 9 | 1010 | 71 | ||
| AK149989.1 | - | 8 - 9 | 963 | 71 | ||
| AK155611.1 | - | 8 - 9 | 879 | 71 | ||
| BC051678.1 | - | 5 - 6 | 536 | 71 | ||
| BC058535.1 | - | 7 - 8 | 692 | 71 | ||
| BC079531.1 | - | 7 - 8 | 705 | 71 | ||
| BC094323.1 | - | 8 - 9 | 1015 | 71 | ||
| BC094423.1 | - | 8 - 9 | 966 | 6812389 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubr7
Genomic Map