Assay Details
| Gene Symbol: Ppip5k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178795.4 | NP_848910.3 | 16 - 17 | 1899 | 68 | |
| GenBank mRNA | AF502585.1 | - | 16 - 17 | 1901 | 68 | |
| AK046696.1 | - | 17 - 18 | 1941 | 68 | ||
| AK083140.1 | - | 16 - 17 | 1910 | 68 | ||
| AK122264.1 | - | 16 - 17 | 1865 | 68 | ||
| BC039977.1 | - | 2 - 3 | 249 | 68 | ||
| BC138351.1 | - | 15 - 16 | 1797 | 8861371 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppip5k1
Genomic Map