Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200013693.mm.1 |
NM_001109761.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013693.mm.1 |
NM_001109761.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013693.mm.1 |
NM_001177799.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013693.mm.1 |
NM_001177799.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013693.mm.1 |
NM_007601.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013693.mm.1 |
NM_007601.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013693.mm.1 |
TC0200005480.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200013694.mm.1 |
NM_001109761.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013694.mm.1 |
NM_001109761.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013694.mm.1 |
NM_001177799.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013694.mm.1 |
NM_001177799.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013694.mm.1 |
NM_007601.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013694.mm.1 |
NM_007601.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013694.mm.1 |
TC0200005480.mm.1 |
MTA TC |