Assay Details
| Gene Symbol: Qsox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK140324.1 | - | 1 - 2 | 349 | 123 | |
| AK157992.1 | - | 1 - 2 | 342 | 123 | ||
| AK165642.1 | - | 1 - 2 | 351 | 123 | ||
| BC030934.1 | - | 1 - 2 | 383 | 4920915 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Qsox2
Genomic Map